Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Mpdu1
Entrez Gene ID
100689049
Gene Name
mannose-P-dolichol utilization defect 1
Gene Aliases
I79_002743, SL15
Chromosome Location
Chr.Un|NW_003613643: 1711841 - 1717040 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001244037.1 NP_001230966.1 1 - 2 119 101
GenBank mRNA AF121896.1 - 1 - 2 119 101
JI886339.1 - 2 - 3 101
U55387.2 - 1 - 2 119 101

View Full Product Details