Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Gna11
Entrez Gene ID
100689065
Gene Name
guanine nucleotide binding protein (G protein), alpha 11 (Gq class)
Gene Aliases
I79_013821
Chromosome Location
Chr.Un|NW_003614244: 352484 - 357164 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001244051.1 NP_001230980.1 1 - 2 323 91
GenBank mRNA DQ202706.1 - 1 - 2 323 91
JI862434.1 - 1 - 2 185 91
JP039111.1 - 1 - 2 186 91

View Full Product Details