Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Pde6g
Entrez Gene ID
100764811
Gene Name
phosphodiesterase 6G, cGMP-specific, rod, gamma
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003613637: 2700724 - 2706236 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003496885.2 XP_003496933.1 3 - 4 439 104

View Full Product Details