Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Nkg7
Entrez Gene ID
100752474
Gene Name
natural killer cell granule protein 7
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003615616: 122537 - 123316 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003512841.1 XP_003512889.1 1 - 2 152 73

View Full Product Details