Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Zcchc24
Entrez Gene ID
100754690
Gene Name
zinc finger, CCHC domain containing 24
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003613953: 1263210 - 1313646 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003503257.2 XP_003503305.1 3 - 4 594 77

View Full Product Details