Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Timm17b
Entrez Gene ID
100762818
Gene Name
translocase of inner mitochondrial membrane 17 homolog B (yeast)
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003614122: 584762 - 594402 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003505098.1 XP_003505146.1 4 - 5 387 75
GenBank mRNA JI875859.1 - 4 - 5 393 75
JP060529.1 - 4 - 5 393 75

View Full Product Details