Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Nnat
Entrez Gene ID
100757948
Gene Name
neuronatin
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003613641: 1384046 - 1388646 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007607849.1 XP_007606039.1 2 - 3 212 57

View Full Product Details