Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Fam71a
Entrez Gene ID
100763531
Gene Name
family with sequence similarity 71, member A
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003615163: 26941 - 28689 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003511579.1 XP_003511627.1 1 - 1 1615 70

View Full Product Details