Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Taf9b
Entrez Gene ID
100767965
Gene Name
TAF9B RNA polymerase II, TATA box binding protein (TBP)-associated factor, 31kDa
Gene Aliases
I79_012488
Chromosome Location
Chr.Un|NW_003614139: 787060 - 794144 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007645822.1 XP_007644012.1 4 - 5 461 89
XM_007645823.1 XP_007644013.1 4 - 5 467 89
GenBank mRNA JI888408.1 - 4 - 5 431 89

View Full Product Details