Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Actl9
Entrez Gene ID
100752462
Gene Name
actin-like 9
Gene Aliases
I79_018803
Chromosome Location
Chr.Un|NW_003614895: 38177 - 39424 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003510497.1 XP_003510545.1 1 - 1 924 89

View Full Product Details