Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Krt73
Entrez Gene ID
100752555
Gene Name
keratin 73
Gene Aliases
I79_021073
Chromosome Location
Chr.Un|NW_003615420: 174603 - 182936 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003512319.2 XP_003512367.2 7 - 8 1331 72

View Full Product Details