Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Nes
Entrez Gene ID
100761741
Gene Name
nestin
Gene Aliases
I79_009594
Chromosome Location
Chr.Un|NW_003613923: 829715 - 851207 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007643548.1 XP_007641738.1 7 - 8 868 78

View Full Product Details