Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Eif2d
Entrez Gene ID
100759038
Gene Name
eukaryotic translation initiation factor 2D
Gene Aliases
I79_007610, Ligatin
Chromosome Location
Chr.Un|NW_003613838: 1735929 - 1752879 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003501218.2 XP_003501266.1 10 - 11 1353 69
XM_007641906.1 XP_007640096.1 10 - 11 1189 69
GenBank mRNA JP061130.1 - 5 - 6 624 69

View Full Product Details