Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Sit1
Entrez Gene ID
100774102
Gene Name
signaling threshold regulating transmembrane adaptor 1
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003614108: 224112 - 225090 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003504990.2 XP_003505038.2 1 - 2 80 61

View Full Product Details