Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Tmem72
Entrez Gene ID
100759477
Gene Name
transmembrane protein 72
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003615739: 76372 - 99715 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003513115.1 XP_003513163.1 1 - 2 76 86

View Full Product Details