Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Prrt4
Entrez Gene ID
100760257
Gene Name
proline-rich transmembrane protein 4
Gene Aliases
I79_003121
Chromosome Location
Chr.Un|NW_003613652: 1342129 - 1351909 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007613790.1 XP_007611980.1 5 - 6 1379 77

View Full Product Details