Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Wdr20
Entrez Gene ID
100772230
Gene Name
WD repeat domain 20
Gene Aliases
I79_004671
Chromosome Location
Chr.Un|NW_003613702: 364922 - 389304 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007633131.1 XP_007631321.1 2 - 3 1472 72

View Full Product Details