Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Nudt14
Entrez Gene ID
100770798
Gene Name
nudix (nucleoside diphosphate linked moiety X)-type motif 14
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003613702: 2501650 - 2508293 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003498747.2 XP_003498795.1 3 - 4 122 98

View Full Product Details