Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Gpr25
Entrez Gene ID
100766966
Gene Name
G protein-coupled receptor 25
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003613706: 2067133 - 2089366 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003498894.1 XP_003498942.1 2 - 3 1174 85

View Full Product Details