Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Adam21
Entrez Gene ID
100763081
Gene Name
ADAM metallopeptidase domain 21
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003613686: 21920 - 24070 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007624975.1 XP_007623165.1 1 - 1 2023 131

View Full Product Details