Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Rsl24d1
Entrez Gene ID
100767126
Gene Name
ribosomal L24 domain containing 1
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003613582: 3284260 - 3294666 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003495020.2 XP_003495068.1 5 - 6 491 85
GenBank mRNA JI886744.1 - 5 - 6 483 85
JP039861.1 - 5 - 6 483 85

View Full Product Details