Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Mrpl40
Entrez Gene ID
100772641
Gene Name
mitochondrial ribosomal protein L40
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003613898: 386898 - 392490 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003502445.2 XP_003502493.1 2 - 3 295 88
GenBank mRNA JI882302.1 - 2 - 3 202 88
JP040152.1 - 2 - 3 202 88

View Full Product Details