Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Slc16a13
Entrez Gene ID
100763738
Gene Name
solute carrier family 16, member 13
Gene Aliases
I79_002779
Chromosome Location
Chr.Un|NW_003613643: 2259637 - 2264619 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007609301.1 XP_007607491.1 2 - 3 201 106

View Full Product Details