Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Tmem27
Entrez Gene ID
100772257
Gene Name
transmembrane protein 27
Gene Aliases
Collectrin, I79_009987
Chromosome Location
Chr.Un|NW_003613952: 52417 - 81720 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003503234.1 XP_003503282.1 4 - 5 313 70

View Full Product Details