Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Psca
Entrez Gene ID
100767033
Gene Name
prostate stem cell antigen
Gene Aliases
I79_020048
Chromosome Location
Chr.Un|NW_003615135: 300231 - 308851 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007652064.1 XP_007650254.1 2 - 3 249 70

View Full Product Details