Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LOC100774181
Entrez Gene ID
100774181
Gene Name
chromosome unknown open reading frame, human C2orf43
Gene Aliases
I79_008571
Chromosome Location
Chr.Un|NW_003613876: 103747 - 157274 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003502097.2 XP_003502145.1 4 - 5 547 78
GenBank mRNA JI876754.1 - 4 - 5 548 78

View Full Product Details