Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Mrpl50
Entrez Gene ID
100755968
Gene Name
mitochondrial ribosomal protein L50
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003614021: 484166 - 491671 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003504019.2 XP_003504067.1 1 - 2 139 60

View Full Product Details