Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Chrna9
Entrez Gene ID
100756671
Gene Name
cholinergic receptor, nicotinic, alpha 9 (neuronal)
Gene Aliases
I79_018066
Chromosome Location
Chr.Un|NW_003614780: 420689 - 434161 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007650504.1 XP_007648694.1 2 - 3 220 73

View Full Product Details