Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Calhm2
Entrez Gene ID
100753368
Gene Name
calcium homeostasis modulator 2
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003615368: 70881 - 77430 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003512140.2 XP_003512188.1 2 - 3 1054 95

View Full Product Details