Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Mkrn3
Entrez Gene ID
100770562
Gene Name
makorin ring finger protein 3
Gene Aliases
I79_016374
Chromosome Location
Chr.Un|NW_003614542: 190689 - 208044 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003508430.2 XP_003508478.1 1 - 2 57 67

View Full Product Details