Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Psmg4
Entrez Gene ID
100758919
Gene Name
proteasome (prosome, macropain) assembly chaperone 4
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003613650: 71933 - 79258 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003497472.2 XP_003497520.1 1 - 2 275 74

View Full Product Details