Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Wfdc12
Entrez Gene ID
100762925
Gene Name
WAP four-disulfide core domain 12
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003614077: 565608 - 565939 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003504590.1 XP_003504638.1 1 - 2 70 67

View Full Product Details