Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Bcl6b
Entrez Gene ID
100764025
Gene Name
B-cell CLL/lymphoma 6, member B
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003613643: 2267880 - 2271267 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003497257.2 XP_003497305.1 5 - 6 921 72

View Full Product Details