Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Hoxc11
Entrez Gene ID
100771213
Gene Name
homeobox C11
Gene Aliases
I79_014806
Chromosome Location
Chr.Un|NW_003614339: 95541 - 97545 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003507215.1 XP_003507263.1 1 - 2 482 75

View Full Product Details