Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Olfm4
Entrez Gene ID
100755691
Gene Name
olfactomedin 4
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003614426: 6327 - 27153 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003507785.1 XP_003507833.1 2 - 3 327 89

View Full Product Details