Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Aph1b
Entrez Gene ID
100772143
Gene Name
APH1B gamma secretase subunit
Gene Aliases
I79_005385
Chromosome Location
Chr.Un|NW_003613730: 1525653 - 1545309 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003499338.2 XP_003499386.1 4 - 5 479 83

View Full Product Details