Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Slc35g1
Entrez Gene ID
100756137
Gene Name
solute carrier family 35, member G1
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003613805: 852714 - 875446 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003500718.2 XP_003500766.1 1 - 2 111 90

View Full Product Details