Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Mcat
Entrez Gene ID
100770744
Gene Name
malonyl CoA:ACP acyltransferase (mitochondrial)
Gene Aliases
I79_015852
Chromosome Location
Chr.Un|NW_003614471: 338922 - 348020 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003508034.2 XP_003508082.1 2 - 3 450 65

View Full Product Details