Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Mal2
Entrez Gene ID
100755368
Gene Name
mal, T-cell differentiation protein 2
Gene Aliases
I79_007338
Chromosome Location
Chr.Un|NW_003613825: 1539891 - 1604544 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007641718.1 XP_007639908.1 3 - 4 568 80

View Full Product Details