Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
B3galtl
Entrez Gene ID
100759864
Gene Name
beta 1,3-galactosyltransferase-like
Gene Aliases
I79_002074
Chromosome Location
Chr.Un|NW_003613630: 220709 - 287267 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007652246.1 XP_007650436.1 3 - 4 221 65
GenBank mRNA JP038488.1 - 3 - 4 321 65

View Full Product Details