Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Dnajc8
Entrez Gene ID
100771757
Gene Name
DnaJ (Hsp40) homolog, subfamily C, member 8
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003613689: 2411670 - 2429872 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003498383.2 XP_003498431.1 8 - 9 671 89
GenBank mRNA JI868499.1 - 8 - 9 673 89
JP060965.1 - 8 - 9 673 89

View Full Product Details