Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Mrpl20
Entrez Gene ID
100771139
Gene Name
mitochondrial ribosomal protein L20
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003614798: 362265 - 369476 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003509973.2 XP_003510021.1 3 - 4 330 66
GenBank mRNA JI865232.1 - 3 - 4 312 66
JP062182.1 - 3 - 4 312 66

View Full Product Details