Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Kif25
Entrez Gene ID
100750864
Gene Name
kinesin family member 25
Gene Aliases
I79_011043
Chromosome Location
Chr.Un|NW_003614028: 781222 - 813551 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003504064.1 XP_003504112.1 5 - 6 631 114

View Full Product Details