Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Fam83f
Entrez Gene ID
100771028
Gene Name
family with sequence similarity 83, member F
Gene Aliases
I79_015146
Chromosome Location
Chr.Un|NW_003614389: 259293 - 288098 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003507468.1 XP_003507516.1 2 - 3 278 67

View Full Product Details