Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Rasl11a
Entrez Gene ID
100758419
Gene Name
RAS-like, family 11, member A
Gene Aliases
I79_002113
Chromosome Location
Chr.Un|NW_003613630: 3097153 - 3099797 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003496661.2 XP_003496709.1 3 - 4 366 70
GenBank mRNA JI874109.1 - 3 - 4 355 70

View Full Product Details