Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Gast
Entrez Gene ID
100772161
Gene Name
gastrin
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003614078: 19678 - 20103 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003504620.1 XP_003504668.1 1 - 2 216 77

View Full Product Details