Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
March11
Entrez Gene ID
100766895
Gene Name
membrane-associated ring finger (C3HC4) 11
Gene Aliases
I79_011777
Chromosome Location
Chr.Un|NW_003614080: 1074467 - 1194297 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003504685.2 XP_003504733.2 2 - 3 150 80

View Full Product Details