Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ttll8
Entrez Gene ID
100754335
Gene Name
tubulin tyrosine ligase-like family, member 8
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003614727: 84298 - 128889 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003509554.1 XP_003509602.1 7 - 8 884 69

View Full Product Details