Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Odf3b
Entrez Gene ID
100759457
Gene Name
outer dense fiber of sperm tails 3B
Gene Aliases
I79_017763
Chromosome Location
Chr.Un|NW_003614727: 528047 - 530092 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003509570.2 XP_003509618.1 1 - 2 110 63

View Full Product Details