Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Gprin3
Entrez Gene ID
100769431
Gene Name
GPRIN family member 3
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003615025: 113671 - 115938 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003511082.1 XP_003511130.1 1 - 1 2125 127

View Full Product Details